MilliporeSigma
  • Absence of donor CD40 protects renal allograft epithelium and preserves renal function.
Absence of donor CD40 protects renal allograft epithelium and preserves renal function.
Transplant international : official journal of the European Society for Organ Transplantation (2013-02-15)
Anna Katharina Kraus, Pietro Ernesto Cippá, Ariana Gaspert, Jin Chen, Ilka Edenhofer, Rudolph Peter Wüthrich, Maja Lindenmeyer, Stephan Segerer, Thomas Fehr
ABSTRACT
Blocking the CD40-CD154 pathway prevents allograft rejection and induces donor-specific tolerance in various experimental models. However, the translation to clinical studies has been hampered by unexpected thromboembolic complications of CD154-blocking antibodies. Thus, blocking CD40 instead is now considered as an alternative strategy. Here, we evaluated the role of donor CD40 in allospecific T-cell responses in vitro and in an in vivo model for renal transplantation. Fully MHC-mismatched allografts from CD40-deficient donors displayed better renal function than wild type. These functional data correlated with a lower level of apoptosis in renal tubular epithelial cells and higher expression of PD-L1, which is most probably because of a reduced Th17 response in recipients of a CD40-deficient donor. This hypothesis was supported in vitro, where donor CD40 expression was important for the induction of direct allospecific T-cell responses. Especially the induction of Th17 cells was critically dependent on donor CD40. IL-17A in conjunction with interferon-γ in turn rendered renal tubular epithelial cells to a more costimulatory state by upregulating CD40 and downregulating PD-L1 expression. In conclusion, CD40 blockade not only reduces the allospecific T-cell responses, but might also lead to protection of tubular epithelium from apoptosis and thereby preserve kidney allograft function.
Sigma-Aldrich
Anti-DNA Antibody, single stranded specific, clone F7-26, clone F7-26, Chemicon®, from mouse