MilliporeSigma
Deoxyfluorination of phenols.
Journal of the American Chemical Society (2011-07-09)
Pingping Tang, Weike Wang, Tobias Ritter
ABSTRACT
An operationally simple ipso fluorination of phenols with a new deoxyfluorination reagent is presented.
Sigma-Aldrich
Phenofluor solution, 0.1 M in toluene
Sigma-Aldrich
2-Chloro-1,3-bis(2,6-diisopropylphenyl)imidazolium chloride, ≥95%