MilliporeSigma
  • Mutant Huntingtin Secretion in Neuro2A Cells and Rat Primary Cortical Neurons.
Mutant Huntingtin Secretion in Neuro2A Cells and Rat Primary Cortical Neurons.
Bio-protocol (2018-01-13)
Katarina Trajkovic, Hyunkyung Jeong, Dimitri Krainc
ABSTRACT
Quantitative analysis of proteins secreted from the cells poses a challenge due to their low abundance and the interfering presence of a large amount of bovine serum albumin (BSA) in the cell culture media. We established assays for detection of mutant huntingtin (mHtt) secreted from Neuro2A cell line stably expressing mHtt and rat primary cortical neurons by Western blotting. Our protocol is based on reducing the amounts of BSA in the media while maintaining cell viability and secretory potential, and concentrating the media prior to analysis by means of ultrafiltration.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture