MilliporeSigma
Proangiogenic Properties of Thrombospondin-4.
Arteriosclerosis, thrombosis, and vascular biology (2015-07-04)
Santoshi Muppala, Ella Frolova, Roy Xiao, Irene Krukovets, Suzy Yoon, George Hoppe, Amit Vasanji, Edward Plow, Olga Stenina-Adognravi
ABSTRACT
Thrombospondin-4 (TSP-4) is 1 of the 5 members of the thrombospondin protein family. TSP-1 and TSP-2 are potent antiangiogenic proteins. However, angiogenic properties of the 3 other TSPs, which do not contain the domains associated with the antiangiogeneic activity of TSP-1 and TSP-2, have not been explored. In our previous studies, we found that TSP-4 is expressed in the vascular matrix of blood vessels of various sizes and is especially abundant in capillaries. We sought to identify the function of TSP-4 in the regulation of angiogenesis. The effect of TSP-4 in in vivo angiogenesis models and its effect on angiogenesis-related properties in cultured cells were assessed using Thbs4(-/-) mice, endothelial cells (EC) derived from these mice, and recombinant TSP-4. Angiogenesis was decreased in Thbs4(-/-) mice compared with wild-type mice. TSP-4 was detected in the lumen of the growing blood vessels. Mice expressing the P387 TSP-4 variant, which was previously associated with coronary artery disease and found to be more active in its cellular interactions, displayed greater angiogenesis compared with A387 form. Lung EC from Thbs4(-/-) mice exhibited decreased adhesion, migration, and proliferation capacities compared with EC from wild-type mice. Recombinant TSP-4 promoted proliferation and the migration of EC. Integrin α2 and gabapentin receptor α2δ-1 were identified as receptors involved in regulation of EC adhesion, migration, and proliferation by TSP-4. TSP-4, an extracellular matrix protein previously associated with tissue remodeling, is now demonstrated to possess proangiogenic activity.
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, Azide Free, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, Azide Free, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone B3A, azide free, clone B3A, Chemicon®, from mouse