MilliporeSigma
  • [Vitamin D metabolism and signaling in human hepatocellular carcinoma and surrounding non-tumorous liver].
[Vitamin D metabolism and signaling in human hepatocellular carcinoma and surrounding non-tumorous liver].
Orvosi hetilap (2016-11-29)
Evelin Horváth, Bernadett Balla, János Kósa, Péter András Lakatos, Áron Lazáry, Dániel Németh, Hasan Jozilan, Áron Somorácz, Anna Korompay, Benedek Gyöngyösi, Katalin Borka, András Kiss, Péter Kupcsulik, Zsuzsa Schaff, Ferenc Szalay
ABSTRACT
1,25-Dihydroxy vitamin D3 mediates antitumor effects in hepatocellular carcinoma. We examined mRNA and protein expression differences in 1,25-Dihydroxy vitamin D3-inactivating CYP24A1, mRNA of activating CYP27B1 enzymes, and that of VDR between human hepatocellular carcinoma and surrounding non-tumorous liver. Snap-frozen tissues from 13 patients were studied for mRNA and protein expression of CYP24A1. Paraffin-embedded tissues from 36 patients were used to study mRNA of VDR and CYP27B1. mRNA expression was measured by RT-PCR, CYP24A1 protein was detected by immunohistochemistry. Expression of VDR and CYP27B1 was significantly lower in hepatocellular carcinoma compared with non-tumorous liver (p<0.05). The majority of the HCC samples expressed CYP24A1 mRNA, but neither of the non-tumorous liver. The gene activation was followed by CYP24A1 protein synthesis. The presence of CYP24A1 mRNA and the reduced expression of VDR and CYP27B1 mRNA in human hepatocellular carcinoma samples indicate decreased bioavailability of 1,25-Dihydroxy vitamin D3, providing an escape mechanism from the anti-tumor effect. Orv. Hetil., 2016, 157(48), 1910-1918.
Sigma-Aldrich
Monoclonal Anti-CYP24A1 antibody produced in mouse, clone 1F8, purified immunoglobulin, buffered aqueous solution