MilliporeSigma
  • IL-31 regulates differentiation and filaggrin expression in human organotypic skin models.
IL-31 regulates differentiation and filaggrin expression in human organotypic skin models.
The Journal of allergy and clinical immunology (2011-12-20)
Christian Cornelissen, Yvonne Marquardt, Katharina Czaja, Jörg Wenzel, Jorge Frank, Juliane Lüscher-Firzlaff, Bernhard Lüscher, Jens M Baron
ABSTRACT
Atopic dermatitis (AD) is an inflammatory skin disease affecting 10% to 20% of children and 1% to 3% of adults in industrialized countries. Enhanced expression of IL-31 is detected in skin samples of patients with AD, but its physiological relevance is not known. We sought to determine the role of IL-31 in skin differentiation. We used human 3-dimensional organotypic skin models with either primary keratinocytes or HaCaT keratinocytes with inducible IL-31 receptor α to evaluate the effect of IL-31. The consequences were studied by using histology, the expression of markers analyzed by immunofluoresence and quantitative RT-PCR, and gene expression arrays. We observed that IL-31 interferes with keratinocyte differentiation. Gene expression analysis revealed a limited set of genes deregulated in response to IL-31, including IL20 and IL24. In HaCaT keratinocytes with inducible IL-31 receptor α, IL-31 inhibited proliferation upon induction of IL-31 receptor α by inducing cell cycle arrest. As in primary cells, IL-31-treated HaCaT cells elicited a differentiation defect in organotypic skin models, associated with reduced epidermal thickness, disturbed epidermal constitution, altered alignment of the stratum basale, and poor development of the stratum granulosum. The differentiation defect was associated with a profound repression of terminal differentiation markers, including filaggrin, an essential factor for skin barrier formation, and a reduced lipid envelope. The highly induced proinflammatory cytokines IL-20 and IL-24 were responsible for part of the effect on FLG expression and thus for terminal differentiation. Our study suggests that IL-31 is an important regulator of keratinocyte differentiation and demonstrates a link between the presence of IL-31 in skin, as found in patients with AD, and filaggrin expression.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-8, clone ASC-8, Chemicon®, from mouse