MilliporeSigma
TGF-β contamination of purified recombinant GDF15.
PloS one (2017-11-22)
Oddrun Elise Olsen, Anette Skjærvik, Berit Fladvad Størdal, Anders Sundan, Toril Holien
ABSTRACT
Purified recombinant proteins for use in biomedical research are invaluable to investigate protein function. However, purity varies in protein batches made in mammalian expression systems, such as CHO-cells or HEK293-cells. This study points to caution while investigating effects of proteins related to the transforming growth factor (TGF)-β superfamily. TGF-β itself is a very potent cytokine and has effects on cells in the femtomolar range. Thus, even very small amounts of contaminating TGF-β in purified protein batches may influence the experimental results given that receptors for TGF-β are present. When we attempted to characterize possible receptors for the TGF-β superfamily ligand GDF15, striking similarities between GDF15-induced activities and known TGF-β activities were found. However, differences between batches of GDF15 were a concern and finally led us to the conclusion that the measured effects were caused by TGF-β and not by GDF15. Our results emphasize that purified recombinant proteins must be used with caution and warrant proper controls. Notably, some conclusions made about GDF15 in already published papers may not be supported by the results shown. Awareness about this issue in the scientific community may prevent spreading of false positive results.
Sigma-Aldrich
Anti-phospho-Smad2 (Ser465/467) Antibody, clone A5S, rabbit monoclonal, culture supernatant, clone A5S, Upstate®