MilliporeSigma
  • Ligand-induced downregulation of TrkA is partly regulated through ubiquitination by Cbl.
Ligand-induced downregulation of TrkA is partly regulated through ubiquitination by Cbl.
FEBS letters (2011-05-17)
Yuga Takahashi, Noriaki Shimokawa, Saeed Esmaeili-Mahani, Akihito Morita, Hiroko Masuda, Toshiharu Iwasaki, Jun'ichi Tamura, Kaisa Haglund, Noriyuki Koibuchi
ABSTRACT
Nerve growth factor (NGF) binding to its receptor TrkA, which belongs to the family of receptor tyrosine kinases (RTKs), is known to induce its internalization, endosomal trafficking and subsequent lysosomal degradation. The Cbl family of ubiquitin ligases plays a major role in mediating ubiquitination and degradation of RTKs. However, it is not known whether Cbl participates in mediating ubiquitination of TrkA. Here we report that c-Cbl mediates ligand-induced ubiquitination and degradation of TrkA. TrkA ubiquitination and degradation required direct interactions between c-Cbl and phosphorylated TrkA. c-Cbl and ubiquitinated TrkA are found in a complex after NGF stimulation and are degraded in lysosomes. Taken together, our data demonstrate that c-Cbl can induce downregulation of NGF-TrkA complexes through ubiquitination and degradation of TrkA.