MilliporeSigma
  • Angiogenesis in differentiated placental multipotent mesenchymal stromal cells is dependent on integrin alpha5beta1.
Angiogenesis in differentiated placental multipotent mesenchymal stromal cells is dependent on integrin alpha5beta1.
PloS one (2009-10-23)
Ming-Yi Lee, Jian-Pei Huang, Yi-Yung Chen, John D Aplin, Yi-Hsin Wu, Chia-Yu Chen, Pei-Chun Chen, Chie-Pein Chen
ABSTRACT
Human placental multipotent mesenchymal stromal cells (hPMSCs) can be isolated from term placenta, but their angiogenic ability and the regulatory pathways involved are not known. hPMSCs were shown to express integrins alpha(v), alpha(4), alpha(5), beta(1), beta(3), and beta(5) and could be induced to differentiate into cells expressing endothelial markers. Increases in cell surface integrins alpha(5) and beta(1), but not alpha(4), alpha(v)beta(3), or alpha(v)beta(5), accompanied endothelial differentiation. Vascular endothelial growth factor-A augmented the effect of fibronectin in enhancing adhesion and migration of differentiated hPMSC through integrin alpha(5)beta(1), but not alpha(v)beta(3) or alpha(v)beta(5). Formation of capillary-like structures in vitro from differentiated cells was inhibited by pre-treatment with function-blocking antibodies to integrins alpha(5) and beta(1). When hPMSCs were seeded onto chick chorioallantoic membranes (CAM), human von Willebrand factor-positive cells were observed to engraft in the chick endothelium. CAMs transplanted with differentiated hPMSCs had a greater number of vessels containing human cells and more incorporated cells per vessel compared to CAMs transplanted with undifferentiated hPMSCs, and overall angiogenesis was enhanced more by the differentiated cells. Function-blocking antibodies to integrins alpha(5) and beta(1) inhibited angiogenesis in the CAM assay. These results suggest that differentiated hPMSCs may contribute to blood vessel formation, and this activity depends on integrin alpha(5)beta(1).
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free, clone HUTS-4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P4G9, ascites fluid, clone P4G9, Chemicon®
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-VE-cadherin Antibody, clone BV6, clone BV6, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse