MilliporeSigma
  • C9orf72-generated poly-GR and poly-PR do not directly interfere with nucleocytoplasmic transport.
C9orf72-generated poly-GR and poly-PR do not directly interfere with nucleocytoplasmic transport.
Scientific reports (2019-11-02)
Joni Vanneste, Thomas Vercruysse, Steven Boeynaems, Adria Sicart, Philip Van Damme, Dirk Daelemans, Ludo Van Den Bosch
ABSTRACT
Repeat expansions in the C9orf72 gene cause amyotrophic lateral sclerosis and frontotemporal dementia characterized by dipeptide-repeat protein (DPR) inclusions. The toxicity associated with two of these DPRs, poly-GR and poly-PR, has been associated with nucleocytoplasmic transport. To investigate the causal role of poly-GR or poly-PR on active nucleocytoplasmic transport, we measured nuclear import and export in poly-GR or poly-PR expressing Hela cells, neuronal-like SH-SY5Y cells and iPSC-derived motor neurons. Our data strongly indicate that poly-GR and poly-PR do not directly impede active nucleocytoplasmic transport.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 4G11, clone 4G11, from mouse
Sigma-Aldrich
Anti-C9ORF72/C9RANT (poly-GR) Antibody, clone 5A2, clone 5A2, 1 mg/mL, from rat