MilliporeSigma
  • Improvements of phosphorescent white OLEDs performance for lighting application.
Improvements of phosphorescent white OLEDs performance for lighting application.
Journal of nanoscience and nanotechnology (2009-02-10)
Jonghee Lee, Hye Yong Chu, Jeong-Ik Lee, Ki-Im Song, Su Jin Lee
ABSTRACT
We developed white OLED device with high power efficiency, in which blue and orange phosphorescent emitters were used. By introduction of multi-functional interlayer which has partial doping of orange dopant inside EBL, we report WOLEDs with peak external efficiencies up to (14.1% EQE, 31.3 Im/W) without light out-coupling technique. At 1000 cd/m2, the performance achieved was 11.9% EQE, 18.7 Im/W with CIE = (0.39, 0.44). We also found that WOLED performances are related with doping ratio of the orange dopant that was inserted inside EBL.