MilliporeSigma
  • Automated untargeted stable isotope assisted lipidomics of liver cells on high glucose shows alteration of sphingolipid kinetics.
Automated untargeted stable isotope assisted lipidomics of liver cells on high glucose shows alteration of sphingolipid kinetics.
Biochimica et biophysica acta. Molecular and cell biology of lipids (2020-02-12)
Davide Noto, Francesca Di Gaudio, Ida Grazia Altieri, Angelo Baldassare Cefalù, Sergio Indelicato, Francesca Fayer, Rossella Spina, Chiara Scrimali, Antonina Giammanco, Alessandro Mattina, Serena Indelicato, Massimiliano Greco, David Bongiorno, Maurizio Averna
ABSTRACT
Untargeted lipidomics is a powerful tool to discover new biomarkers and to understand the physiology and pathology of lipids. The use of stable isotopes as tracers to investigate the kinetics of lipids is another tool able to supply dynamic information on lipid synthesis and catabolism. Coupling the two methodology is then very appealing in the study of lipid metabolism. The main issue to face is to perform thousands of calculations in order to obtain kinetic parameters starting from the MS raw data. An automated computerized routine able to do accomplish such task is presented in this paper. We analyzed the lipid kinetics of palmitic acid (PA) in hepatoma liver cells cultured in vitro in which insulin resistance has been induced by high glucose supplementation. The deuterated palmitate tracer (d5PA) was administered as a bolus and the cells were harvested daily for 48 h. 5dPA was incorporated into 326 monoisotopic compounds and in 84 of their [M + 1] isotopologues detected by high resolution orbitrap MS. The differences between the kinetics curves showed that at least four long chain triglycerides (TG) species incorporated more PA in glucose treated cells, while phosphocholines, sphingomyelins, mono- and di-glycerides and ceramides (Cer) incorporated less tracer under glucose treatment. Nevertheless, Cer amount was increased by glucose treatment. In conclusion we developed an automated powerful algorithm able to model simultaneously hundreds of kinetic curves obtained in a cell culture spiked with a stable isotope tracer, and to analyze the difference between the two different cell models.
Product Description
Avanti
C17 Ceramide (d18:1/17:0), Avanti Research - A Croda Brand
Sigma-Aldrich
Palmitic acid-15,15,16,16,16-d5, 98 atom % D, 99% (CP)