MilliporeSigma
  • Effect of inhaled terbutaline on substrate utilization and 300-kcal time trial performance.
Effect of inhaled terbutaline on substrate utilization and 300-kcal time trial performance.
Journal of applied physiology (Bethesda, Md. : 1985) (2014-09-27)
Anders Kalsen, Morten Hostrup, Sebastian Karlsson, Peter Hemmersbach, Jens Bangsbo, Vibeke Backer
ABSTRACT
In a randomized, double-blind crossover design, we investigated the effect of the beta2-agonist terbutaline (TER) on endurance performance and substrate utilization in nine moderately trained men [maximum oxygen uptake (V̇O(2 max)) 58.9 ± 3.1 ml·min(-1)·kg(-1)]. Subjects performed 60 min of submaximal exercise (65-70% of V̇O(2 max)) immediately followed by a 300-kcal time trial with inhalation of either 15 mg of TER or placebo (PLA). Pulmonary gas exchange was measured during the submaximal exercise, and muscle biopsies were collected before and after the exercise bouts. Time trial performance was not different between TER and PLA (1,072 ± 145 vs. 1,054 ± 125 s). During the submaximal exercise, respiratory exchange ratio, glycogen breakdown (TER 266 ± 32, PLA 195 ± 28 mmol/kg dw), and muscle lactate accumulation (TER 20.3 ± 1.6, PLA 13.2 ± 1.2 mmol/kg dw) were higher (P < 0.05) with TER than PLA. There was no difference between TER and PLA in net muscle glycogen utilization or lactate accumulation during the time trial. Intramyocellular triacylglycerol content did not change with treatment or exercise. Pyruvate dehydrogenase-E1α phosphorylation at Ser(293) and Ser(300) was lower (P < 0.05) before submaximal exercise with TER than PLA, with no difference after the submaximal exercise and the time trial. Before submaximal exercise, acetyl-CoA carboxylase 2 (ACC2) phosphorylation at Ser(221) was higher (P < 0.05) with TER than PLA. There was no difference in phosphorylation of alpha 5'-AMP-activated protein kinase (αAMPK) at Thr(172) between treatments. The present study suggests that beta2-agonists do not enhance 300-kcal time trial performance, but they increase carbohydrate metabolism in skeletal muscles during submaximal exercise independent of AMPK and ACC phosphorylation, and that this effect diminishes as drug exposure time, exercise duration, and intensity are increased.
Sigma-Aldrich
Anti-phospho PDHE1-A type I (Ser300) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography