MilliporeSigma
  • Mechanism of activation of plant monogalactosyldiacylglycerol synthase 1 (MGD1) by phosphatidylglycerol.
Mechanism of activation of plant monogalactosyldiacylglycerol synthase 1 (MGD1) by phosphatidylglycerol.
Glycobiology (2020-02-27)
Milène Nitenberg, Olga Makshakova, Joana Rocha, Serge Perez, Eric Maréchal, Maryse A Block, Agnès Girard-Egrot, Christelle Breton
ABSTRACT
Mono- and digalactosyldiacylglycerol are essential galactolipids for the biogenesis of plastids and functioning of the photosynthetic machinery. In Arabidopsis, the first step of galactolipid synthesis is catalyzed by monogalactosyldiacylglycerol synthase 1 (MGD1), a monotopic protein located in the inner envelope membrane of chloroplasts, which transfers a galactose residue from UDP-galactose to diacylglycerol (DAG). MGD1 needs anionic lipids such as phosphatidylglycerol (PG) to be active, but the mechanism by which PG activates MGD1 is still unknown. Recent studies shed light on the catalytic mechanism of MGD1 and on the possible PG binding site. Particularly, Pro189 was identified as a potential residue implicated in PG binding and His155 as the putative catalytic residue. In the present study, using a multifaceted approach (Langmuir membrane models, atomic force microscopy, molecular dynamics; MD), we investigated the membrane binding properties of native MGD1 and mutants (P189A and H115A). We demonstrated that both residues are involved in PG binding, thus suggesting the existence of a PG-His catalytic dyad that should facilitate deprotonation of the nucleophile hydroxyl group of DAG acceptor. Interestingly, MD simulations showed that MGD1 induces a reorganization of lipids by attracting DAG molecules to create an optimal platform for binding.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Avanti
18:1 DG, 1-2-dioleoyl-sn-glycerol, chloroform
Avanti
18:1 DG, Avanti Research - A Croda Brand