MilliporeSigma
  • The prion protein regulates glutamate-mediated Ca2+ entry and mitochondrial Ca2+ accumulation in neurons.
The prion protein regulates glutamate-mediated Ca2+ entry and mitochondrial Ca2+ accumulation in neurons.
Journal of cell science (2017-07-14)
Agnese De Mario, Caterina Peggion, Maria Lina Massimino, Francesca Viviani, Angela Castellani, Marta Giacomello, Dmitry Lim, Alessandro Bertoli, Maria Catia Sorgato
ABSTRACT
The cellular prion protein (PrPC) whose conformational misfolding leads to the production of deadly prions, has a still-unclarified cellular function despite decades of intensive research. Following our recent finding that PrPC limits Ca2+ entry via store-operated Ca2+ channels in neurons, we investigated whether the protein could also control the activity of ionotropic glutamate receptors (iGluRs). To this end, we compared local Ca2+ movements in primary cerebellar granule neurons and cortical neurons transduced with genetically encoded Ca2+ probes and expressing, or not expressing, PrPC Our investigation demonstrated that PrPC downregulates Ca2+ entry through each specific agonist-stimulated iGluR and after stimulation by glutamate. We found that, although PrP-knockout (KO) mitochondria were displaced from the plasma membrane, glutamate addition resulted in a higher mitochondrial Ca2+ uptake in PrP-KO neurons than in their PrPC-expressing counterpart. This was because the increased Ca2+ entry through iGluRs in PrP-KO neurons led to a parallel increase in Ca2+-induced Ca2+ release via ryanodine receptor channels. These data thus suggest that PrPC takes part in the cell apparatus controlling Ca2+ homeostasis, and that PrPC is involved in protecting neurons from toxic Ca2+ overloads.
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Metabotropic Glutamate Receptor 5 Antibody, pain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 831, Chemicon®, from rabbit