MilliporeSigma
  • Viral DNA-Dependent Induction of Innate Immune Response to Hepatitis B Virus in Immortalized Mouse Hepatocytes.
Viral DNA-Dependent Induction of Innate Immune Response to Hepatitis B Virus in Immortalized Mouse Hepatocytes.
Journal of virology (2015-10-23)
Xiuji Cui, Daniel N Clark, Kuancheng Liu, Xiao-Dong Xu, Ju-Tao Guo, Jianming Hu
ABSTRACT
Hepatitis B virus (HBV) infects hundreds of millions of people worldwide and causes acute and chronic hepatitis, cirrhosis, and hepatocellular carcinoma. HBV is an enveloped virus with a relaxed circular (RC) DNA genome. In the nuclei of infected human hepatocytes, conversion of RC DNA from the incoming virion or cytoplasmic mature nucleocapsid (NC) to the covalently closed circular (CCC) DNA, which serves as the template for producing all viral transcripts, is essential to establish and sustain viral replication. A prerequisite for CCC DNA formation is the uncoating (disassembly) of NCs to expose their RC DNA content for conversion to CCC DNA. We report here that in an immortalized mouse hepatocyte cell line, AML12HBV10, in which RC DNA exposure is enhanced, the exposed viral DNA could trigger an innate immune response that was able to modulate viral gene expression and replication. When viral gene expression and replication were low, the innate response initially stimulated these processes but subsequently acted to shut off viral gene expression and replication after they reached peak levels. Inhibition of viral DNA synthesis or cellular DNA sensing and innate immune signaling diminished the innate response. These results indicate that HBV DNA, when exposed in the host cell cytoplasm, can function to trigger an innate immune response that, in turn, modulates viral gene expression and replication. Chronic infection by hepatitis B virus (HBV) afflicts hundreds of millions worldwide and is sustained by the episomal covalently closed circular (CCC) DNA in the nuclei of infected hepatocytes. Release of viral genomic DNA from cytoplasmic nucleocapsids (NCs) (NC disassembly or uncoating) is a prerequisite for its conversion to CCC DNA, which can also potentially expose the viral DNA to host DNA sensors and trigger an innate immune response. We have found that in an immortalized mouse hepatocyte cell line in which efficient CCC DNA formation was associated with enhanced exposure of nucleocapsid-associated DNA, the exposed viral DNA indeed triggered host cytoplasmic DNA sensing and an innate immune response that was able to modulate HBV gene expression and replication. Thus, HBV can, under select conditions, be recognized by the host innate immune response through exposed viral DNA, which may be exploited therapeutically to clear viral persistence.
Sigma-Aldrich
Anti-cGAS Antibody, from rabbit, purified by affinity chromatography