MilliporeSigma
  • Flow Cytometry-Based Detection and Analysis of BCL-2 Family Proteins and Mitochondrial Outer Membrane Permeabilization (MOMP).
Flow Cytometry-Based Detection and Analysis of BCL-2 Family Proteins and Mitochondrial Outer Membrane Permeabilization (MOMP).
Methods in molecular biology (Clifton, N.J.) (2018-12-12)
Lindsey M Ludwig, Katrina L Maxcy, James L LaBelle
ABSTRACT
The BCL-2 family of proteins orchestrates a complex signaling network that governs the balance between cellular survival and death. A comprehensive understanding of the mechanistic interactions between these proteins continues to evolve in normal and malignant cells. The functional variation by individual BCL-2 proteins in different cell types has driven clinical therapeutic development in targeting individual BCL-2 members with the goal of fine-tuning cell death in diseased cells. Given the importance of understanding and validating the effect of activating or inhibiting BCL-2 protein interactions in individual cells, the methods used to measure apoptotic cell death have undergone increased scrutiny. Here, we describe two in vitro flow cytometry-based methods that are useful in measuring BCL-2 proteins and mitochondrial-based cell death in complex cell populations.
Sigma-Aldrich
Anti-Bak (NT) Antibody, from rabbit, purified by affinity chromatography