MilliporeSigma
  • Ligand-independent antiapoptotic function of estrogen receptor-beta in lung cancer cells.
Ligand-independent antiapoptotic function of estrogen receptor-beta in lung cancer cells.
Molecular endocrinology (Baltimore, Md.) (2010-07-28)
Guangfeng Zhang, Naveena Yanamala, Kira L Lathrop, Lin Zhang, Judith Klein-Seetharaman, Harish Srinivas
ABSTRACT
Recent studies have demonstrated the presence of estrogen receptor (ER)beta in the mitochondria in various cell types and tissues, but the exact function of this localization remains unclear. In this study, we have examined the function of mitochondrial ERbeta in non-small-cell lung cancer (NSCLC) cells. Down-regulation of ERbeta by short hairpin RNA constructs sensitized NSCLC cells to various apoptosis-inducing agents such as cisplatin, taxol, and etoposide. The increased growth inhibition and induction of apoptosis in ERbeta-knockdown cells was observed irrespective of estrogen treatment, suggesting a ligand-independent role of ERbeta in regulating the intrinsic apoptotic pathway. Further, ERbeta from the mitochondrial fraction physically interacted with the proapoptotic protein Bad, in a ligand-independent manner. Glutathione-S-transferase pull-down assays and molecular modeling studies revealed that the DNA-binding domain and hinge region of ERbeta, and the BH3 domain of Bad were involved in these interactions. Further investigations revealed that ERbeta inhibited Bad function by disrupting Bad-Bcl-X(L) and Bad-Bcl-2 interactions. Reintroduction of ERbeta in the mitochondria of ERbeta knockdown cells reversed their sensitivity to cisplatin. Overall, our results demonstrate a ligand-independent role of ERbeta in regulating apoptosis, revealing a novel function for ERbeta in the mitochondria.
Sigma-Aldrich
Anti-Estrogen Receptor β Antibody, clone 68-4, rabbit monoclonal, clone 68-4, Upstate®, from rabbit