MilliporeSigma
  • PTK7+ Mononuclear Cells Express VEGFR2 and Contribute to Vascular Stabilization by Upregulating Angiopoietin-1.
PTK7+ Mononuclear Cells Express VEGFR2 and Contribute to Vascular Stabilization by Upregulating Angiopoietin-1.
Arteriosclerosis, thrombosis, and vascular biology (2015-05-23)
Sunil K Chauhan, Hyung Keun Lee, Hyun Soo Lee, Eun Young Park, Eunae Jeong, Reza Dana
ABSTRACT
In angiogenesis, circulating mononuclear cells are recruited to vascular lesions; however, the underlying mechanisms are poorly understood. Here, we characterize the functional role of protein tyrosine kinase 7 (PTK7)-expressing CD11b(+) mononuclear cells in vitro and in vivo using a mouse model of angiogenesis. Although the frequencies of PTK7(+)CD11b(+) cells in the bone marrow remained similar after vascular endothelial growth factor-A-induced neovascularization, we observed an 11-fold increase in the cornea. Importantly, vascular endothelial growth factor-A-induced chemotaxis of PTK7(+) cells was mediated by vascular endothelial growth factor receptor 2. In a coculture with endothelial cells, PTK7(+)CD11b(+) cells stabilized the vascular network for 2 weeks by expressing high levels of angiopoietin-1. The enhanced vascular stability was abolished by knockdown of angiopoietin-1 in PTK7(+)CD11b(+) cells and could be restored by angiopoietin-1 treatment. We conclude that PTK7 expression in perivascular mononuclear cells induces vascular endothelial growth factor receptor 2 and angiopoietin-1 expression and thus contributes to vascular stabilization in angiogenesis.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Anti-Angiopoietin-1 Antibody, NT, Chemicon®, from rabbit
Sigma-Aldrich
SN50 trifluoroacetate salt, ≥95% (HPLC)
Sigma-Aldrich
Anti-Angiopoietin-2 Antibody, NT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-phospho-Tie2 (Ser1119) Antibody, from rabbit, purified by affinity chromatography