MilliporeSigma
  • Generation of an induced pluripotent stem cell line (TRNDi002-B) from a patient carrying compound heterozygous p.Q208X and p.G310G mutations in the NGLY1 gene.
Generation of an induced pluripotent stem cell line (TRNDi002-B) from a patient carrying compound heterozygous p.Q208X and p.G310G mutations in the NGLY1 gene.
Stem cell research (2019-01-07)
Rong Li, Manisha Pradhan, Miao Xu, Amanda Baskfield, Atena Farkhondeh, Yu-Shan Cheng, Jeanette Beers, Jizhong Zou, Chengyu Liu, Matthew Might, Steven Rodems, Wei Zheng
ABSTRACT
NGLY1 deficiency is a rare genetic disease caused by mutations in the NGLY1 gene that encodes N-glycanase 1. The disease phenotype in patient cells is unclear. A human induced pluripotent stem cell (iPSC) line was generated from skin dermal fibroblasts of a patient with NGLY1 deficiency that has compound heterozygous mutations of a p.Q208X variant (c.622C > T) in exon 4 and a p.G310G variant (c.930C > T) in exon 6 of the NGLY1 gene. This iPSC line offers a useful resource to study the disease pathophysiology and a cell-based model for drug development to treat NGLY1 deficiency.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit