MilliporeSigma
  • Selective degradation of host MicroRNAs by an intergenic HCMV noncoding RNA accelerates virus production.
Selective degradation of host MicroRNAs by an intergenic HCMV noncoding RNA accelerates virus production.
Cell host & microbe (2013-06-19)
Sanghyun Lee, Jaewon Song, Sungchul Kim, Jongkyu Kim, Yujin Hong, Youngkyun Kim, Donghyun Kim, Daehyun Baek, Kwangseog Ahn
ABSTRACT
Virulence of human cytomegalovirus (HCMV) clinical isolates correlates with carriage of a 15 kb segment in the UL/b' region of the viral genome, which is absent from attenuated strains. The mechanisms by which this segment contributes to HCMV virulence remain obscure. We observed that intergenic RNA sequences within the 15 kb segment function as a microRNA (miRNA) decay element (miRDE) and direct the selective, sequence-specific turnover of mature miR-17 and miR-20a encoded within the host miR-17-92 cluster. Unlike canonical miRNA-mRNA interactions, the miRNA-miRDE interactions did not repress miRDE expression. miRNA binding site mutations retargeted miRDE to other miR-17-92 cluster miRNAs, which are otherwise resistant to miRDE-mediated decay. miRDE function was required to accelerate virus production in the context of lytic HCMV infection. These results indicate a role for viral noncoding RNA in regulating cellular miRNAs during HCMV pathogenesis and suggest that noncoding RNAs may play a role in mature miRNA turnover.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Anti-Cytomegalovirus Antibody, clone 8B1.2, clone 8B1.2, Chemicon®, from mouse
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture