MilliporeSigma
  • A simple high-throughput approach identifies actionable drug sensitivities in patient-derived tumor organoids.
A simple high-throughput approach identifies actionable drug sensitivities in patient-derived tumor organoids.
Communications biology (2019-03-02)
Nhan Phan, Jenny J Hong, Bobby Tofig, Matthew Mapua, David Elashoff, Neda A Moatamed, Jin Huang, Sanaz Memarzadeh, Robert Damoiseaux, Alice Soragni
ABSTRACT
Tumor organoids maintain cell-cell interactions, heterogeneity, microenvironment, and drug response of the sample they originate from. Thus, there is increasing interest in developing tumor organoid models for drug development and personalized medicine applications. Although organoids are in principle amenable to high-throughput screenings, progress has been hampered by technical constraints and extensive manipulations required by current methods. Here we introduce a miniaturized method that uses a simplified geometry by seeding cells around the rim of the wells (mini-rings). This allows high-throughput screenings in a format compatible with automation as shown using four patient-derived tumor organoids established from two ovarian and one peritoneal high-grade serous carcinomas and one carcinosarcoma of the ovary. Using our automated screening platform, we identified personalized responses by measuring viability, number, and size of organoids after exposure to 240 kinase inhibitors. Results are available within a week from surgery, a timeline compatible with therapeutic decision-making.
Sigma-Aldrich
Anti-CDK7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution