MilliporeSigma
  • Lymphocyte Responses to Chymotrypsin- or Trypsin V-Digested beta-Lactoglobulin in Patients with Cow's Milk Allergy.
Lymphocyte Responses to Chymotrypsin- or Trypsin V-Digested beta-Lactoglobulin in Patients with Cow's Milk Allergy.
Allergy, asthma, and clinical immunology : official journal of the Canadian Society of Allergy and Clinical Immunology (2007-03-15)
Masashi Kondo, Toshiyuki Fukao, Shinji Shinoda, Norio Kawamoto, Hideo Kaneko, Zenichiro Kato, Eiko Matsui, Takahide Teramoto, Taku Nakano, Naomi Kondo
ABSTRACT
: Chymotrypsin- or trypsin V- (a mixture of trypsin and chymotrypsin) digested beta-lactoglobulin (BLG) peptides were prepared and were confirmed to have much less immunoglobulin (lg)G and lgE reactivity compared with intact BLG by IgG inhibition enzymelinked immunosorbent assay and IgE dot blotting. The lymphocyte responses to intact BLG and these peptides were examined using peripheral blood mononuclear cells (PBMCs) from 10 patients with cow's milk allergy. The PBMCs from most patients had lower lymphocyte responses to chymotrypsin- and trypsin V-digested BLG peptides than those to intact BLG. However, PBMCs from one and two patients retained significant proliferative responses to both peptides and to only the former peptide, respectively. Interferon-c production stimulated by chymotrypsin-digested peptides was still detectable in all five patients tested. Chymotrypsindigested BLG reduced lgE reactivity but still induced some lymphocyte responses.
Sigma-Aldrich
Anti-Human IgE−Alkaline Phosphatase antibody, Mouse monoclonal, clone GE-1, purified from hybridoma cell culture