MilliporeSigma
  • Resistin is expressed in human macrophages and directly regulated by PPAR gamma activators.
Resistin is expressed in human macrophages and directly regulated by PPAR gamma activators.
Biochemical and biophysical research communications (2002-12-31)
Lisa Patel, Amy C Buckels, Ian J Kinghorn, Paul R Murdock, Joanna D Holbrook, Christopher Plumpton, Colin H Macphee, Stephen A Smith
ABSTRACT
Resistin is a cysteine-rich protein postulated to be a molecular link between obesity and type 2 diabetes. The aim of this study was to investigate the role of PPAR gamma in the regulation of resistin expression in human primary macrophages. Fluorescent real-time PCR (Taqman) analysis of resistin expression across a range of human tissues showed that resistin is highly expressed in bone marrow compared to other tissues. Taqman analysis and Western blotting showed that rosiglitazone decreased resistin expression at both the mRNA and protein levels in human primary monocyte-derived macrophages in vitro. Resistin expression was reduced by up to 80% after exposure to 100 nM rosiglitazone for 96 h. Bioinformatics analysis of the genomic sequence upstream of the resistin coding sequence identified several putative PPAR response elements of which one was shown to bind PPAR gamma using electrophoretic mobility shift assays. Our data support a direct role for PPAR gamma in the regulation of resistin expression.
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution