MilliporeSigma
  • Planar cell polarity protein Dishevelled 3 (Dvl3) regulates ectoplasmic specialization (ES) dynamics in the testis through changes in cytoskeletal organization.
Planar cell polarity protein Dishevelled 3 (Dvl3) regulates ectoplasmic specialization (ES) dynamics in the testis through changes in cytoskeletal organization.
Cell death & disease (2019-02-28)
Linxi Li, Baiping Mao, Ming Yan, Siwen Wu, Renshan Ge, Qingquan Lian, C Yan Cheng
ABSTRACT
In the mammalian testes, such as in rats, the directional alignment of polarized elongating/elongated spermatids, in particular step 17-19 spermatids, across the plane of seminiferous epithelium resembles planar cell polarity (PCP) found in hair cells of the cochlea. It is obvious that spermatid PCP is necessary to support the simultaneous development of maximal number of elongating/elongated spermatids to sustain the daily production of > 50 million sperm per adult rat. Studies have shown that the testis indeed expresses multiple PCP proteins necessary to support spermatid PCP. Herein, using physiological and biochemical assays, and morphological analysis, and with the technique of RNA interference (RNAi) to knockdown PCP protein Dishevelled (Dvl) 1 (Dvl1), Dvl2, Dvl3, or Dvl1/2/3, Dvl proteins, in particular Dvl3, it was shown that Dvl3 played a crucial role of support Sertoli cell tight junction (TJ)-permeability barrier function through changes in the organization of actin- and microtubule (MT)-based cytoskeletons. More important, an in vivo knockdown of Dvl1/2/3 in the testis, defects of spermatid polarity were remarkably noted across the seminiferous epithelium, concomitant with defects of spermatid adhesion and spermatid transport, leading to considerably defects in spermatogenesis. More important, Dvl1/2/3 triple knockdown in the testis also impeded the organization of actin- and MT-based cytoskeletons owing to disruptive spatial expression of actin- and MT-regulatory proteins. In summary, PCP Dishevelled proteins, in particular, Dvl3 is a regulator of Sertoli cell blood-testis barrier (BTB)  and also spermatid PCP function through its effects on the actin- and MT-based cytoskeletons in Sertoli cells.
Sigma-Aldrich
Monoclonal Anti-DVL3 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ARP3 antibody, Mouse monoclonal, clone FMS338, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid