MilliporeSigma
  • Effect of riboflavin on the photolysis of folic acid in aqueous solution.
Effect of riboflavin on the photolysis of folic acid in aqueous solution.
Journal of pharmaceutical and biomedical analysis (2000-11-30)
M J Akhtar, M A Khan, I Ahmad
ABSTRACT
A study of the photolysis of folic acid in aqueous solution by visible radiation in the presence of riboflavin has been made. The second-order rate constants for the bimolecular interaction of folic acid and riboflavin have been determined in the pH range 4.0-9.0. The rate pH profile shows a gradual increase in the rate up to pH 6.2 (pHmax) followed by a decrease up to pH 9.0, depending upon the susceptibility of ionic species involved in the interaction. The rate of photolysis varies from 0.50 x 10(3) M(-1) min(-1) (pH 9.0) and 0.63 x 10(3) M(-1) min(-1) (pH 4.0) to 3.0 x 10(3) M(-1) min(-1) (pH 6.2) in the pH range studied. A HPLC method has been used for the assay of folic acid and its photoproducts, pterine-6-carboxylic acid and p-aminobenzoyl-L-glutamic acid in the presence of riboflavin.
Sigma-Aldrich
Pterine-6-carboxylic acid, ≥98.0% (HPLC)