MilliporeSigma
  • Defective granulation tissue formation in mice with specific ablation of integrin-linked kinase in fibroblasts - role of TGFβ1 levels and RhoA activity.
Defective granulation tissue formation in mice with specific ablation of integrin-linked kinase in fibroblasts - role of TGFβ1 levels and RhoA activity.
Journal of cell science (2010-10-29)
Katrin Blumbach, Manon C Zweers, Georg Brunner, Andreas S Peters, Markus Schmitz, Jan-Niklas Schulz, Alexander Schild, Christopher P Denton, Takao Sakai, Reinhard Fässler, Thomas Krieg, Beate Eckes
ABSTRACT
Wound healing crucially relies on the mechanical activity of fibroblasts responding to TGFβ1 and to forces transmitted across focal adhesions. Integrin-linked kinase (ILK) is a central adapter recruited to integrin β1 tails in focal adhesions mediating the communication between cells and extracellular matrix. Here, we show that fibroblast-restricted inactivation of ILK in mice leads to impaired healing due to a severe reduction in the number of myofibroblasts, whereas inflammatory infiltrate and vascularization of the granulation tissue are unaffected. Primary ILK-deficient fibroblasts exhibit severely reduced levels of extracellular TGFβ1, α-smooth muscle actin (αSMA) production and myofibroblast conversion, which are rescued by exogenous TGFβ1. They are further characterized by elevated RhoA and low Rac1 activities, resulting in abnormal shape and reduced directional migration. Interference with RhoA-ROCK signaling largely restores morphology, migration and TGFβ1 levels. We conclude that, in fibroblasts, ILK is crucial for limiting RhoA activity, thus promoting TGFβ1 production, which is essential for dermal repair following injury.
Sigma-Aldrich
MS-SAFE Protease and Phosphatase Inhibitor
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Vinculin Antibody, clone VIIF9 (7F9), clone VIIF9 (7F9), Chemicon®, from mouse