MilliporeSigma
  • Enzymatic properties of mouse 25-hydroxyvitamin D3 1 alpha-hydroxylase expressed in Escherichia coli.
Enzymatic properties of mouse 25-hydroxyvitamin D3 1 alpha-hydroxylase expressed in Escherichia coli.
European journal of biochemistry (1999-03-27)
T Sakaki, N Sawada, K Takeyama, S Kato, K Inouye
ABSTRACT
Renal 25-hydroxyvitamin D3 1 alpha-hydroxylase cDNA cloned from the kidneys of mice lacking the vitamin D receptor was expressed in Escherichia coli JM109. As expected, the bacterially-expressed enzyme catalyzes the 1 alpha-hydroxylation of 25-hydroxyvitamin D3 with a Michaelis constant, K(m), value of 2.7 microM. Unexpectedly, the enzyme also hydroxylates the 1 alpha-position of 24,25-dihydroxyvitamin D3 with a K(m) of 1.3 microM, and a fourfold higher Vmax/K(m) compared with the 25-hydroxyvitamin D3 hydroxylase activity, suggesting that 24,25-dihydroxyvitamin D3 is a better substrate than 25-hydroxyvitamin D3 for 1 alpha-hydroxylase. In addition, the enzyme showed 1 alpha-hydroxylase activity toward 24-oxo-25-hydroxyvitamin D3. However, it showed only slight activity towards 23,25-dihydroxyvitamin D3 and 24-oxo-23,25-dihydroxyvitamin D3, and no detectable activity towards vitamin D3 and 24,25,26,27-tetranor-23-hydroxyvitamin D3. These results suggest that the 25-hydroxyl group of vitamin D3 is essential for the 1 alpha-hydroxylase activity and the 24-hydroxyl group enhances the activity, but the 23-hydroxyl group greatly reduced the activity. Another remarkable finding is that living recombinant E. coli cells can convert the substrates into the 1 alpha-hydroxylated products, suggesting the presence of a redox partner of 1 alpha-hydroxylase in E. coli cells.
Sigma-Aldrich
(24R)-24,25-Dihydroxyvitamin D3, ≥98% (vitamin + pre-vitamin, HPLC)