MilliporeSigma
  • Natural Killer Cells Degenerate Intact Sensory Afferents following Nerve Injury.
Natural Killer Cells Degenerate Intact Sensory Afferents following Nerve Injury.
Cell (2019-02-05)
Alexander J Davies, Hyoung Woo Kim, Rafael Gonzalez-Cano, Jahyang Choi, Seung Keun Back, Seung Eon Roh, Errin Johnson, Melanie Gabriac, Mi-Sun Kim, Jaehee Lee, Jeong Eun Lee, Yun Sook Kim, Yong Chul Bae, Sang Jeong Kim, Kyung-Mi Lee, Heung Sik Na, Priscilla Riva, Alban Latremoliere, Simon Rinaldi, Sophie Ugolini, Michael Costigan, Seog Bae Oh
ABSTRACT
Sensory axons degenerate following separation from their cell body, but partial injury to peripheral nerves may leave the integrity of damaged axons preserved. We show that an endogenous ligand for the natural killer (NK) cell receptor NKG2D, Retinoic Acid Early 1 (RAE1), is re-expressed in adult dorsal root ganglion neurons following peripheral nerve injury, triggering selective degeneration of injured axons. Infiltration of cytotoxic NK cells into the sciatic nerve by extravasation occurs within 3 days following crush injury. Using a combination of genetic cell ablation and cytokine-antibody complex stimulation, we show that NK cell function correlates with loss of sensation due to degeneration of injured afferents and reduced incidence of post-injury hypersensitivity. This neuro-immune mechanism of selective NK cell-mediated degeneration of damaged but intact sensory axons complements Wallerian degeneration and suggests the therapeutic potential of modulating NK cell function to resolve painful neuropathy through the clearance of partially damaged nerves.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography