MilliporeSigma
Pulmonary surfactant: functions and molecular composition.
Biochimica et biophysica acta (1998-11-14)
J Goerke
ABSTRACT
This review briefly notes recent findings important for understanding the surface mechanical functions of pulmonary surfactant. Currently known surfactant-specific proteins and lipids are discussed, with an eye to their possible functions. Competing models of the alveolar subphase life cycle of surfactant are also presented. It is concluded that, in spite of much effort, we still do not understand the basic molecular mechanisms underlying surfactant's rapid adsorption to the air-water interface.