MilliporeSigma
  • Inhibition of motor neuron death in vitro and in vivo by a p75 neurotrophin receptor intracellular domain fragment.
Inhibition of motor neuron death in vitro and in vivo by a p75 neurotrophin receptor intracellular domain fragment.
Journal of cell science (2015-10-28)
Dusan Matusica, Fabienne Alfonsi, Bradley J Turner, Tim J Butler, Stephanie R Shepheard, Mary-Louise Rogers, Sune Skeldal, Clare K Underwood, Marie Mangelsdorf, Elizabeth J Coulson
ABSTRACT
The p75 neurotrophin receptor (p75(NTR); also known as NGFR) can mediate neuronal apoptosis in disease or following trauma, and facilitate survival through interactions with Trk receptors. Here we tested the ability of a p75(NTR)-derived trophic cell-permeable peptide, c29, to inhibit p75(NTR)-mediated motor neuron death. Acute c29 application to axotomized motor neuron axons decreased cell death, and systemic c29 treatment of SOD1(G93A) mice, a common model of amyotrophic lateral sclerosis, resulted in increased spinal motor neuron survival mid-disease as well as delayed disease onset. Coincident with this, c29 treatment of these mice reduced the production of p75(NTR) cleavage products. Although c29 treatment inhibited mature- and pro-nerve-growth-factor-induced death of cultured motor neurons, and these ligands induced the cleavage of p75(NTR) in motor-neuron-like NSC-34 cells, there was no direct effect of c29 on p75(NTR) cleavage. Rather, c29 promoted motor neuron survival in vitro by enhancing the activation of TrkB-dependent signaling pathways, provided that low levels of brain-derived neurotrophic factor (BDNF) were present, an effect that was replicated in vivo in SOD1(G93A) mice. We conclude that the c29 peptide facilitates BDNF-dependent survival of motor neurons in vitro and in vivo.
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat