MilliporeSigma
  • Immunohistochemical validation of COL3A1, GPR158 and PITHD1 as prognostic biomarkers in early-stage ovarian carcinomas.
Immunohistochemical validation of COL3A1, GPR158 and PITHD1 as prognostic biomarkers in early-stage ovarian carcinomas.
BMC cancer (2019-09-20)
Hanna Engqvist, Toshima Z Parris, Anikó Kovács, Szilárd Nemes, Elisabeth Werner Rönnerman, Shahin De Lara, Jana Biermann, Karin Sundfeldt, Per Karlsson, Khalil Helou
ABSTRACT
Ovarian cancer is the main cause of gynecological cancer-associated death. However, 5-year survival rates differ dramatically between the five main ovarian carcinoma histotypes. Therefore, we need to have a better understanding of the mechanisms that promote histotype-specific ovarian carcinogenesis and identify novel prognostic biomarkers. Here, we evaluated the prognostic role of 29 genes for early-stage (I and II) ovarian carcinomas (n = 206) using immunohistochemistry (IHC). We provide evidence of aberrant protein expression patterns for Collagen type III alpha 1 chain (COL3A1), G protein-coupled receptor 158 (GPR158) and PITH domain containing 1 (PITHD1). Kaplan-Meier survival analysis revealed that COL3A1 expression was associated with shorter overall survival in the four major histotypes of epithelial ovarian carcinoma patients (P value = 0.026, HR = 2.99 (95% CI 1.089-8.19)). Furthermore, GPR158 and PITHD1 were shown to be histotype-specific prognostic biomarkers, with elevated GPR158 expression patterns in mucinous ovarian carcinoma patients with unfavorable overall survival (P value = 0.00043, HR = 6.13 (95% CI 1.98-18.98)), and an association with lower PITHD1 protein expression and unfavorable overall and disease-specific survival in clear-cell ovarian carcinoma patients (P value = 0.012, HR = 0.22 (95% CI 0.058-0.80); P value = 0.003, HR = 0.17 (95% CI 0.043-0.64)). The novel biomarkers identified here may improve prognostication at the time of diagnosis and may assist in the development of future individualized therapeutic strategies for ovarian carcinoma patients.
Sigma-Aldrich
Anti-C7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-COL3A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TRIO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-IGHG1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-KIF26B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-CES3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SLC9A4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARMC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GPR158 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MUC15 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP7D2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TRIM71 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EHD3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-JCHAIN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CDH18 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution