MilliporeSigma
  • Exosomal microRNA predicts and protects against severe bronchopulmonary dysplasia in extremely premature infants.
Exosomal microRNA predicts and protects against severe bronchopulmonary dysplasia in extremely premature infants.
JCI insight (2018-03-09)
Charitharth Vivek Lal, Nelida Olave, Colm Travers, Gabriel Rezonzew, Kalsang Dolma, Alexandra Simpson, Brian Halloran, Zubair Aghai, Pragnya Das, Nirmal Sharma, Xin Xu, Kristopher Genschmer, Derek Russell, Tomasz Szul, Nengjun Yi, J Edwin Blalock, Amit Gaggar, Vineet Bhandari, Namasivayam Ambalavanan
ABSTRACT
Premature infants are at high risk for developing bronchopulmonary dysplasia (BPD), characterized by chronic inflammation and inhibition of lung development, which we have recently identified as being modulated by microRNAs (miRNAs) and alterations in the airway microbiome. Exosomes and exosomal miRNAs may regulate cell differentiation and tissue and organ development. We discovered that tracheal aspirates from infants with severe BPD had increased numbers of, but smaller, exosomes compared with term controls. Similarly, bronchoalveolar lavage fluid from hyperoxia-exposed mice (an animal model of BPD) and supernatants from hyperoxia-exposed human bronchial epithelial cells (in vitro model of BPD) had increased exosomes compared with air controls. Next, in a prospective cohort study of tracheal aspirates obtained at birth from extremely preterm infants, utilizing independent discovery and validation cohorts, we identified unbiased exosomal miRNA signatures predictive of severe BPD. The strongest signal of reduced miR-876-3p in BPD-susceptible compared with BPD-resistant infants was confirmed in the animal model and in vitro models of BPD. In addition, based on our recent discovery of increased Proteobacteria in the airway microbiome being associated with BPD, we developed potentially novel in vivo and in vitro models for BPD combining Proteobacterial LPS and hyperoxia exposure. Addition of LPS led to a larger reduction in exosomal miR 876-3p in both hyperoxia and normoxia compared with hyperoxia alone, thus indicating a potential mechanism by which alterations in microbiota can suppress miR 876-3p. Gain of function of miR 876-3p improved the alveolar architecture in the in vivo BPD model, demonstrating a causal link between miR 876-3p and BPD. In summary, we provide evidence for the strong predictive biomarker potential of miR 876-3p in severe BPD. We also provide insights on the pathogenesis of neonatal lung disease, as modulated by hyperoxia and microbial product-induced changes in exosomal miRNA 876-3p, which could be targeted for future therapeutic development.
Sigma-Aldrich
Anti-CEACAM8 antibody produced in rabbit, affinity isolated antibody