MilliporeSigma
Opposing Roles of apolipoprotein E in aging and neurodegeneration.
Life science alliance (2019-02-15)
Eloise Hudry, Jacob Klickstein, Claudia Cannavo, Rosemary Jackson, Alona Muzikansky, Sheetal Gandhi, David Urick, Taylie Sargent, Lauren Wrobleski, Allyson D Roe, Steven S Hou, Kishore V Kuchibhotla, Rebecca A Betensky, Tara Spires-Jones, Bradley T Hyman
ABSTRACT
Apolipoprotein E (APOE) effects on brain function remain controversial. Removal of APOE not only impairs cognitive functions but also reduces neuritic amyloid plaques in mouse models of Alzheimer's disease (AD). Can APOE simultaneously protect and impair neural circuits? Here, we dissociated the role of APOE in AD versus aging to determine its effects on neuronal function and synaptic integrity. Using two-photon calcium imaging in awake mice to record visually evoked responses, we found that genetic removal of APOE improved neuronal responses in adult APP/PSEN1 mice (8-10 mo). These animals also exhibited fewer neuritic plaques with less surrounding synapse loss, fewer neuritic dystrophies, and reactive glia. Surprisingly, the lack of APOE in aged mice (18-20 mo), even in the absence of amyloid, disrupted visually evoked responses. These results suggest a dissociation in APOE's role in AD versus aging: APOE may be neurotoxic during early stages of amyloid deposition, although being neuroprotective in latter stages of aging.
Sigma-Aldrich
Anti-β-Amyloid antibody, Mouse monoclonal, clone BAM-10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Anti-NR2A Antibody, clone A12W, rabbit monoclonal, culture supernatant, clone A12W, Upstate®
Sigma-Aldrich
Anti-Synapsin I Antibody, Chemicon®, from rabbit