MilliporeSigma
  • Human macular Müller cells rely more on serine biosynthesis to combat oxidative stress than those from the periphery.
Human macular Müller cells rely more on serine biosynthesis to combat oxidative stress than those from the periphery.
eLife (2019-05-01)
Ting Zhang, Ling Zhu, Michele C Madigan, Wei Liu, Weiyong Shen, Svetlana Cherepanoff, Fanfan Zhou, Shaoxue Zeng, Jianhai Du, Mark C Gillies
ABSTRACT
The human macula is more susceptible than the peripheral retina to developing blinding conditions such as age-related macular degeneration, diabetic retinopathy. A key difference between them may be the nature of their Müller cells. We found primary cultured Müller cells from macula and peripheral retina display significant morphological and transcriptomic differences. Macular Müller cells expressed more phosphoglycerate dehydrogenase (PHGDH, a rate-limiting enzyme in serine synthesis) than peripheral Müller cells. The serine synthesis, glycolytic and mitochondrial function were more activated in macular than peripheral Müller cells. Serine biosynthesis is critical in defending against oxidative stress. Intracellular reactive oxygen species and glutathione levels were increased in primary cultured macular Müller cells which were more susceptible to oxidative stress after inhibition of PHGDH. Our findings indicate serine biosynthesis is a critical part of the macular defence against oxidative stress and suggest dysregulation of this pathway as a potential cause of macular pathology.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-3-PGDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
CBR-5884, ≥98% (HPLC)