MilliporeSigma
  • Interaction of FUS and HDAC1 regulates DNA damage response and repair in neurons.
Interaction of FUS and HDAC1 regulates DNA damage response and repair in neurons.
Nature neuroscience (2013-09-17)
Wen-Yuan Wang, Ling Pan, Susan C Su, Emma J Quinn, Megumi Sasaki, Jessica C Jimenez, Ian R A Mackenzie, Eric J Huang, Li-Huei Tsai
ABSTRACT
Defects in DNA repair have been extensively linked to neurodegenerative diseases, but the exact mechanisms remain poorly understood. We found that FUS, an RNA/DNA-binding protein that has been linked to amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration, is important for the DNA damage response (DDR). The function of FUS in DDR involved a direct interaction with histone deacetylase 1 (HDAC1), and the recruitment of FUS to double-stranded break sites was important for proper DDR signaling. Notably, FUS proteins carrying familial ALS mutations were defective in DDR and DNA repair and showed a diminished interaction with HDAC1. Moreover, we observed increased DNA damage in human ALS patients harboring FUS mutations. Our findings suggest that an impaired DDR and DNA repair may contribute to the pathogenesis of neurodegenerative diseases linked to FUS mutations.
Sigma-Aldrich
Anti-BRCA2 Antibody, clone 5.23, clone 5.23, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse