MilliporeSigma
  • What information could the main actors of liquid biopsy provide? -a representative case of non-small cell lung cancer (NSCLC).
What information could the main actors of liquid biopsy provide? -a representative case of non-small cell lung cancer (NSCLC).
Journal of thoracic disease (2018-09-04)
Aldo Pezzuto, Mariangela Manicone, Maria Chiara Scaini, Alberto Ricci, Salvatore Mariotta, Rita Zamarchi, Elisabetta Rossi
ABSTRACT
In non-small cell lung cancer (NSCLC), there is a consensus regarding the use of liquid biopsy, generally, to detect "druggable" mutations and, in particular, to monitor tyrosine kinase inhibitor (TKI) treatments. However, whether circulating tumor cells (CTCs) are better tools than cell-free DNA (cfDNA), is still a matter of debate, mainly concerning which antigen(s) we should use to investigating simultaneously both epithelial and epithelial-to-mesenchymal transient (EMT) phenotype in the same sample of CTCs. To address this item, we exploited here a single-tube liquid biopsy, to detect both epithelial cell adhesion molecule (EpCAM)-positive CTCs and EpCAM-low/negative CTCs, because down-modulation of EpCAM is considered the first step in EMT. Furthermore, we analyzed the DNA from CTCs of four different phenotypes (ctcDNA), according to their EpCAM expression and cytokeratin pattern, and circulating tumor DNA (ctDNA) by droplet digital PCR (ddPCR), in order to disclose activating and resistance-driving mutations. Liquid biopsy reflected spatial and temporal heterogeneity of the tumor under treatment pressure. We provide the proof-of-concept that the complementary use of ctDNA and ctcDNA represents a reliable, minimally invasive and dynamic tool for a more comprehensive view of tumor evolution.
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone LL002, FITC conjugated, clone LL002, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated, clone LP5K, Chemicon®, from mouse