MilliporeSigma
  • TP-064, a potent and selective small molecule inhibitor of PRMT4 for multiple myeloma.
TP-064, a potent and selective small molecule inhibitor of PRMT4 for multiple myeloma.
Oncotarget (2018-05-03)
Kazuhide Nakayama, Magdalena M Szewczyk, Carlo Dela Sena, Hong Wu, Aiping Dong, Hong Zeng, Fengling Li, Renato Ferreira de Freitas, Mohammad S Eram, Matthieu Schapira, Yuji Baba, Mihoko Kunitomo, Douglas R Cary, Michiko Tawada, Akihiro Ohashi, Yasuhiro Imaeda, Kumar Singh Saikatendu, Charles E Grimshaw, Masoud Vedadi, Cheryl H Arrowsmith, Dalia Barsyte-Lovejoy, Atsushi Kiba, Daisuke Tomita, Peter J Brown
ABSTRACT
Protein arginine methyltransferase (PRMT) 4 (also known as coactivator-associated arginine methyltransferase 1; CARM1) is involved in a variety of biological processes and is considered as a candidate oncogene owing to its overexpression in several types of cancer. Selective PRMT4 inhibitors are useful tools for clarifying the molecular events regulated by PRMT4 and for validating PRMT4 as a therapeutic target. Here, we report the discovery of TP-064, a potent, selective, and cell-active chemical probe of human PRMT4 and its co-crystal structure with PRMT4. TP-064 inhibited the methyltransferase activity of PRMT4 with high potency (half-maximal inhibitory concentration, IC50 < 10 nM) and selectivity over other PRMT family proteins, and reduced arginine dimethylation of the PRMT4 substrates BRG1-associated factor 155 (BAF155; IC50= 340 ± 30 nM) and Mediator complex subunit 12 (MED12; IC50 = 43 ± 10 nM). TP-064 treatment inhibited the proliferation of a subset of multiple myeloma cell lines, with affected cells arrested in G1 phase of the cell cycle. TP-064 and its negative control (TP-064N) will be valuable tools to further investigate the biology of PRMT4 and the therapeutic potential of PRMT4 inhibition.
Sigma-Aldrich
Anti-dimethyl-BAF155 Antibody (Arg1064, asymetrically di-methylated), from rabbit, purified by affinity chromatography