MilliporeSigma
  • β-catenin inhibits promyelocytic leukemia protein tumor suppressor function in colorectal cancer cells.
β-catenin inhibits promyelocytic leukemia protein tumor suppressor function in colorectal cancer cells.
Gastroenterology (2011-12-14)
Reiko Satow, Miki Shitashige, Takafumi Jigami, Kiyoko Fukami, Kazufumi Honda, Issay Kitabayashi, Tesshi Yamada
ABSTRACT
Loss of promyelocytic leukemia protein (PML) nuclear body (NB) formation has been reported in colorectal and other solid tumors. However, genetic alteration of PML is rarely observed in these tumors; the exact mechanisms that mediate loss of PML function are not known. We previously used a comprehensive shotgun mass spectrometry approach to identify PML as 1 of 70 proteins that coimmunoprecipitate with anti-T-cell factor 4 in DLD-1 and HCT116 colorectal cancer cell lines; we investigated the effects of altered β-catenin expression on PML function in these cells. β-catenin specifically interacted with the product of PML transcript variant IV (PML-IV) through the armadillo repeat domain of β-catenin. Overexpression of β-catenin in colorectal cancer cells disrupted the subcellular compartmentalization of PML-IV, whereas knockdown of β-catenin restored formation of PML-NB. Modification of PML by the small ubiquitin-related modifier (SUMO) is required for proper assembly of PML-NB. β-catenin inhibited Ran-binding protein 2-mediated SUMOylation of PML-IV. β-catenin interacts with PML isoform IV and disrupts PML-IV function and PML-NB formation by inhibiting Ran-binding protein 2-mediated SUMO modification of PML-IV. These findings indicate the involvement of a posttranslational mechanism in disruption of PML-NB organization in cancer cells and provide more information about the oncogenic functions of β-catenin.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TCF-4 Antibody, clone 6H5-3, clone 6H5-3, Upstate®, from mouse
Sigma-Aldrich
Anti-Glutathione: N-ethylmaleimide adduct Antibody, clone 8.1GSH, clone 8.1GSH, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid