MilliporeSigma
  • Candidate molecular pathways of white matter vulnerability in the brain of normal aging rhesus monkeys.
Candidate molecular pathways of white matter vulnerability in the brain of normal aging rhesus monkeys.
GeroScience (2018-01-23)
Amy A Robinson, Carmela R Abraham, Douglas L Rosene
ABSTRACT
Mammalian aging is associated with decline in cognitive functions. Studies searching for a cause of cognitive aging initially focused on neuronal loss but quantitative investigations of rat, monkey, and human brain using stereology demonstrated that in normal aging, unlike in neurodegenerative disease, neurons are not lost. Instead, electron microscopic and MRI studies in normal aging monkeys revealed age-related damage to myelin sheaths, loss of axons, and reduction in white matter volume which correlates with cognitive impairments. However, little is known about the cause of myelin defects or associated axon loss. The present study investigates the effect of age on signaling pathways between oligodendroglia and neurons using a custom PCR array to assess the expression of 87 genes of interest in cortical gray matter and white matter from the inferior parietal lobe (IPL) of normal rhesus monkeys ranging in age from 4.2 to 30.4 years old. From this array data, five target genes of interest were selected for further analysis to confirm gene expression and measure protein expression. The most interesting target gene identified is brain-derived neurotrophic factor (BDNF), which was the only gene that was altered at both mRNA and protein levels. In gray matter, BDNF mRNA was decreased. While the level of the mature form of the protein was unchanged, there was a specific decrease in the precursor form of BDNF. These alterations in the BDNF in gray matter could contribute to the vulnerability and loss of the axons with age.
Sigma-Aldrich
Anti-Myelin Oligodendrocyte Glycoprotein (MOG) Antibody, clone 8-18C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Trk B Antibody, pain, a.a. 54-67 rTrkB, serum, Chemicon®