MilliporeSigma
  • Dual-specificity tyrosine phosphorylation-regulated kinase 1A ameliorates insulin resistance in neurons by up-regulating IRS-1 expression.
Dual-specificity tyrosine phosphorylation-regulated kinase 1A ameliorates insulin resistance in neurons by up-regulating IRS-1 expression.
The Journal of biological chemistry (2019-11-15)
Shijiao Tian, Wenming Jia, Mei Lu, Juan Zhao, Xiulian Sun
ABSTRACT
Insulin resistance in the brain is a pathological mechanism that is shared between Alzheimer's disease (AD) and type 2 diabetes mellitus (T2DM). Although aberrant expression and phosphorylation of insulin receptor substrate 1 (IRS-1) contribute to insulin resistance, the underlying mechanism remains elusive. In this study, we used several approaches, including adeno-associated virus-based protein overexpression, immunoblotting, immunoprecipitation, immunohistochemistry, and in situ proximal ligation assays, to investigate the function of dual-specificity tyrosine phosphorylation-regulated kinase 1A (DYRK1A) in IRS-1 regulation and the downstream insulin signaling in neurons. We found that DYRK1A overexpression up-regulated IRS-1 expression by slowing turnover of the IRS-1 protein. We further observed that DYRK1A directly interacted with IRS-1 and phosphorylated IRS-1's multiple serine residues. Of note, DYRK1A and IRS-1 were coordinately up-regulated in the prefrontal cortex of db/db mice brain. Furthermore, DYRK1A overexpression ameliorated chronic high insulin-induced insulin resistance in SH-SY5Y cells as well as in primary rat neurons. These findings suggest that DYRK1A protects against insulin resistance in the brain by elevating IRS-1 expression.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-DYRK1A antibody produced in mouse, clone 7D10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phosphoserine Antibody, clone 4A4, clone 4A4, Upstate®, from mouse
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit