MilliporeSigma
  • Changes in the Neuronal Control of the Urinary Bladder in a Model of Radiation Cystitis.
Changes in the Neuronal Control of the Urinary Bladder in a Model of Radiation Cystitis.
The Journal of pharmacology and experimental therapeutics (2018-03-14)
Daniel Giglio, Lucie Podmolíková, Gunnar Tobin
ABSTRACT
Currently, we have assessed the neuronal control of the urinary bladder in radiation cystitis and whether interstitial cells contribute to the condition. Fourteen days after bladder irradiation (20 Gy), rats were sedated and the urinary bladder was cut into two sagittal halves where electrical field stimulation (EFS; 5-20 Hz) was applied on the pelvic nerve afferents or stretch (80 mN) on one-half of the bladder, while contractions were registered on the contralateral half of the bladder in the absence and presence of increasing doses of imatinib (1-10 mg/kg; inhibitor of c-kit-positive interstitial cells), atropine (1 mg/kg; to block muscarinic M3 receptors), or pyridoxalphosphate-6-azophenyl-2',4'-disulfonic acid (2 mg/kg; P2X1 purinoceptor antagonist). Urinary bladders were also excised for organ bath experiments, Western blot, quantitative polymerase chain reaction, and immunohistochemistry. In vivo, EFS contractions were enhanced after irradiation, and imatinib (1-10 mg/mg) inhibited contractions by EFS and stretched dose-dependently in controls but not in irradiated bladders. In the irradiated bladder in vitro, atropine resistance was increased and imatinib (100 µM) inhibited contractions by EFS and agonists (ATP, methacholine) in irradiated bladders and controls. The urinary bladder expressions of P2X1 purinoceptors, muscarinic M3 receptor, choline acetyltransferase, c-kit, and the agonist of c-kit, stem cell factor, were not changed by irradiation. In conclusion, bladder irradiation affects several levels of neuronal control of the urinary bladder. Interstitial cells may contribute to some of the symptoms associated with radiation cystitis.
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m3 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-P2X1 Purinergic Receptor antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-C-KIT antibody produced in rabbit, affinity isolated antibody