MilliporeSigma
  • Functional characterization of tektin-1 in motile cilia and evidence for TEKT1 as a new candidate gene for motile ciliopathies.
Functional characterization of tektin-1 in motile cilia and evidence for TEKT1 as a new candidate gene for motile ciliopathies.
Human molecular genetics (2017-11-10)
Rebecca Ryan, Marion Failler, Madeline Louise Reilly, Meriem Garfa-Traore, Marion Delous, Emilie Filhol, Thérèse Reboul, Christine Bole-Feysot, Patrick Nitschké, Véronique Baudouin, Serge Amselem, Estelle Escudier, Marie Legendre, Alexandre Benmerah, Sophie Saunier
ABSTRACT
A child presenting with Mainzer-Saldino syndrome (MZSDS), characterized by renal, retinal and skeletal involvements, was also diagnosed with lung infections and airway ciliary dyskinesia. These manifestations suggested dysfunction of both primary and motile cilia, respectively. Targeted exome sequencing identified biallelic mutations in WDR19, encoding an IFT-A subunit previously associated with MZSDS-related chondrodysplasia, Jeune asphyxiating thoracic dysplasia and cranioectodermal dysplasia, linked to primary cilia dysfunction, and in TEKT1 which encodes tektin-1 an uncharacterized member of the tektin family, mutations of which may cause ciliary dyskinesia. Tektin-1 localizes at the centrosome in cycling cells, at basal bodies of both primary and motile cilia and to the axoneme of motile cilia in airway cells. The identified mutations impaired these localizations. In addition, airway cells from the affected individual showed severe motility defects without major ultrastructural changes. Knockdown of tekt1 in zebrafish resulted in phenotypes consistent with a function for tektin-1 in ciliary motility, which was confirmed by live imaging. Finally, experiments in the zebrafish also revealed a synergistic effect of tekt1 and wdr19. Altogether, our data show genetic interactions between WDR19 and TEKT1 likely contributing to the overall clinical phenotype observed in the affected individual and provide strong evidence for TEKT1 as a new candidate gene for primary ciliary dyskinesia.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-RSPH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TEKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse