MilliporeSigma
  • Genotoxic stress causes the accumulation of DNA-dependent protein kinase catalytic subunit phosphorylated at serine 2056 at nuclear speckles and alters pre-mRNA alternative splicing.
Genotoxic stress causes the accumulation of DNA-dependent protein kinase catalytic subunit phosphorylated at serine 2056 at nuclear speckles and alters pre-mRNA alternative splicing.
FEBS open bio (2019-02-15)
Shuang Liu, Yuan Shao, Qi Wang, Yonggong Zhai, Xialu Li
ABSTRACT
RNA splicing has emerged as a critical player in the DNA damage response (DDR). However, the underlying mechanism(s) by which pre-mRNA splicing is coordinately regulated by genotoxic stress has remained largely unclear. Here, we show that a DDR factor, DNA-dependent protein kinase (DNA-PK), participates in the modulation of pre-mRNA splicing in the presence of DNA double-strand break (DSB)-induced genotoxic stress. Through indirect immunostaining, we made the surprising discovery that DNA-PK catalytic subunits (DNA-PKcs) autophosphorylated at serine 2056 (S2056) accumulate at nuclear speckles (dynamic nuclear structures that are enriched with splicing factors), following their dissociation from DSB lesions. Inactivation of DNA-PKcs, either using a small molecule inhibitor or by RNA interference, alters alternative splicing of a set of pre-mRNAs in A549 cells treated with the topoisomerase II inhibitor mitoxantrone, indicative of an involvement of DNA-PKcs in modulating pre-mRNA splicing following genotoxic stress. These findings indicate a novel physical and functional connection between the DNA damage response and pre-mRNA splicing, and enhance our understanding of how mRNA splicing is involved in the cellular response to DSB lesions.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP13, clone BP13, Chemicon®, from mouse