MilliporeSigma
  • Myeloperoxidase influences the complement regulatory function of modified C-reactive protein.
Myeloperoxidase influences the complement regulatory function of modified C-reactive protein.
Innate immunity (2013-11-06)
Peng-cheng Xu, Zhi-ying Li, Xiao-wei Yang, Ming-hui Zhao, Min Chen
ABSTRACT
In patients with active anti-neutrophil cytoplasmic Ab (ANCA)-associated vasculitis (AAV), there are high levels of circulating C-reactive protein (CRP), which can inhibit the alternative complement pathway by binding factor H and triggering the classical complement pathway by binding C1q. However, the alternative, not the classical, complement pathway has been proven to play an important role in AAV. We found that both purified myeloperoxidase (MPO) and MPO released from ANCA-stimulated neutrophils could bind modified CRP (mCRP), but not pentameric CRP. Furthermore, MPO could block the binding between mCRP and factor H, as well as the binding between mCRP and C1q. Binding with mCRP did not influence the enzymatic activity of MPO. Binding with mCRP also did not influence the binding between MPO and its physical inhibitor, ceruloplasmin, as well as the binding between MPO and MPO-ANCA. The results indicated that MPO might be a complement regulator and inhibit the negative regulatory effect of CRP on the alternative complement pathway.
Sigma-Aldrich
Anti-Human IgG−Alkaline Phosphatase antibody, Mouse monoclonal, clone GG-5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-C-Reactive Protein antibody produced in mouse, clone CRP-8, ascites fluid
Sigma-Aldrich
Anti-CRP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-Factor H Goat pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution