MilliporeSigma
  • Metabolically Derived Lysine Acylations and Neighboring Modifications Tune the Binding of the BET Bromodomains to Histone H4.
Metabolically Derived Lysine Acylations and Neighboring Modifications Tune the Binding of the BET Bromodomains to Histone H4.
Biochemistry (2017-09-26)
Michael D Olp, Nan Zhu, Brian C Smith
ABSTRACT
Recent proteomic studies discovered histone lysines are modified by acylations beyond acetylation. These acylations derive from acyl-CoA metabolites, potentially linking metabolism to transcription. Bromodomains bind lysine acylation on histones and other nuclear proteins to influence transcription. However, the extent bromodomains bind non-acetyl acylations is largely unknown. Also unclear are the effects of neighboring post-translational modifications, especially within heavily modified histone tails. Using peptide arrays, binding assays, sucrose gradients, and computational methods, we quantified 10 distinct acylations for binding to the bromodomain and extraterminal domain (BET) family. Four of these acylations (hydroxyisobutyrylation, malonylation, glutarylation, and homocitrullination) had never been tested for bromodomain binding. We found N-terminal BET bromodomains bound acetylated and propionylated peptides, consistent with previous studies. Interestingly, all other acylations inhibited binding of the BET bromodomains to peptides and nucleosomes. To understand how context tunes bromodomain binding, effects of neighboring methylation, phosphorylation, and acylation within histone H4 tails were determined. Serine 1 phosphorylation inhibited binding of the BRD4 N-terminal bromodomain to polyacetylated H4 tails by >5-fold, whereas methylation had no effect. Furthermore, binding of BRDT and BRD4 N-terminal bromodomains to H4K5acetyl was enhanced 1.4-9.5-fold by any neighboring acylation of lysine 8, indicating a secondary H4K8acyl binding site that is more permissive of non-acetyl acylations than previously appreciated. In contrast, C-terminal BET bromodomains exhibited 9.9-13.5-fold weaker binding for polyacylated than for monoacylated H4 tails, indicating the C-terminal bromodomains do not cooperatively bind multiple acylations. These results suggest acyl-CoA levels tune or block recruitment of the BET bromodomains to histones, linking metabolism to bromodomain-mediated transcription.
Sigma-Aldrich
Anti-Histidine Tagged Antibody, clone HIS.H8, clone HIS.H8, Upstate®, from mouse