MilliporeSigma
Kras Is Critical for B Cell Lymphopoiesis.
Journal of immunology (Baltimore, Md. : 1950) (2016-01-17)
Yuhong Chen, Yongwei Zheng, Xiaona You, Mei Yu, Guoping Fu, Xinlin Su, Fen Zhou, Wen Zhu, Zhihong Wu, Jing Zhang, Renren Wen, Demin Wang
ABSTRACT
The three major Ras members, Kras, Hras, and Nras, are highly homologous and individual Ras genes can have distinct biological functions. Embryonic lethality of Kras-deficient mice precludes study of the biological functions of this Ras family member. In this study, we generated and examined mice with hematopoietic-specific deletion of Kras and bone marrow (BM) chimeric mice with B cell-specific targeted deletion of Kras. Hematopoietic-specific deletion of Kras impaired early B cell development at the pre-B cell stage and late B cell maturation, resulting in the reduction of BM pre-, immature, and mature B cells and peripheral follicular, marginal zone, and B1 mature B cells. In contrast, Kras deficiency did not affect T cell development. Studies of BM chimeric mice with B cell-specific deletion of Kras demonstrated that Kras deficiency intrinsically impaired B cell development. Kras deficiency reduced BCR-induced B cell proliferation and survival. Furthermore, Kras deficiency specifically impaired pre-BCR- and BCR-induced activation of the Raf-1/MEK/ERK pathway in pre-B and mature B cells, respectively. Thus, Kras is the unique Ras family member that plays a critical role in early B cell development and late B cell maturation through controlling the Raf-1/MEK/ERK pathway.
Sigma-Aldrich
Anti-Ras Antibody, clone RAS10, clone RAS10, Upstate®, from mouse
Sigma-Aldrich
Anti-Fas Antibody (human, neutralizing), clone ZB4, clone ZB4, Upstate®, from mouse