MilliporeSigma
  • GlcNAc Conjugated Atorvastatin with Enhanced Water Solubility and Cellular Internalization.
GlcNAc Conjugated Atorvastatin with Enhanced Water Solubility and Cellular Internalization.
Bioconjugate chemistry (2017-07-27)
Xinfu Zhang, Xiaofang Chen, Weiyu Zhao, Chunxi Zeng, Xiao Luo, Wenqing Li, Bin Li, Justin Jiang, Yizhou Dong
ABSTRACT
Targeting ligands facilitate cell specific drug delivery and improve pharmaceutical properties. Herein, we designed two ligand drug conjugates by conjugating GlcNAc (N-acetylglucosamine) with atorvastatin. These two conjugates, termed G-AT and G-K-AT, exhibited enhanced water solubility and cellular uptake. Moreover, both G-AT and G-K-AT were able to release atorvastatin and consequently achieve significant inhibition against 3-hydroxy-3-methyl-glutaryl-coenzyme A (HMG-CoA) reductase.
Sigma-Aldrich
Anti-LDL Receptor Antibody, clone 2H7.1, clone 2H7.1, from mouse