MilliporeSigma
  • Fermitin family homolog-2 (FERMT2) is highly expressed in human placental villi and modulates trophoblast invasion.
Fermitin family homolog-2 (FERMT2) is highly expressed in human placental villi and modulates trophoblast invasion.
BMC developmental biology (2018-11-02)
Eiko Kawamura, Gina B Hamilton, Ewa I Miskiewicz, Daniel J MacPhee
ABSTRACT
Integrins are transmembrane receptors that mediate cell-extracellular matrix (ECM) and cell-cell adhesion and trophoblast cells undergo changes in integrin expression as they differentiate. However, the mechanism(s) of integrin activation leading to integrin-mediated signaling in trophoblast cell differentiation is unknown. The Fermitin family proteins are integrin activators that help mediate integrin-mediated signaling, but have never been studied in detail within the human placenta. Thus, we examined the spatiotemporal pattern of expression of Fermitin family homolog-2 (FERMT2) in human chorionic villi throughout gestation and its role in trophoblast-substrate adhesion and invasion. Placental villous tissue was obtained from patients undergoing elective terminations by dilatation and curettage at weeks 8-12 (n = 10), weeks 13-14 (n = 8), as well as from term deliveries at weeks 37-40 (n = 6). Tissues were fixed, processed and sections utilized for immunofluorescence analysis of FERMT2 expression during gestation. Additionally, HTR8-SVneo human trophoblast cells were transfected by electroporation with FERMT2-specific siRNAs or non-targeting siRNAs (control) and used in cell-substrate adhesion as well as invasion assays. FERMT2 was more commonly expressed in the basal domain of villous cytotrophoblast cells and prominently localized around the periphery of individual extravillous trophoblast cells. siRNA-mediated knockdown of FERMT2 in HTR8-SVneo cells resulted in significantly decreased trophoblast-substrate attachment (p < 0.05) as well as significantly decreased trophoblast invasion (p < 0.05) relative to control cells. The detection of FERMT2 throughout extravillous trophoblast columns and the results of invasion assays demonstrated that this protein is likely an important regulator of integrin activation in extravillous cells to modulate migration and invasion.
Sigma-Aldrich
Anti-Kindlin-1 antibody, Mouse monoclonal, clone KN-4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ITGA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Integrin α5 Antibody, CT, Intracellular, serum, Chemicon®
Sigma-Aldrich
Anti-von Willebrand Factor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Kindlin-2 Antibody, clone 3A3, clone 3A3, from mouse
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), fluorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.