MilliporeSigma
  • Epigenetic regulation of estrogen receptor alpha gene expression in the mouse cortex during early postnatal development.
Epigenetic regulation of estrogen receptor alpha gene expression in the mouse cortex during early postnatal development.
Endocrinology (2009-12-08)
Jenne M Westberry, Amanda L Trout, Melinda E Wilson
ABSTRACT
Estrogens play a critical role in brain development by acting on areas that express estrogen receptors. In the rodent cortex, estrogen receptor alpha (ER alpha) mRNA expression is high early in postnatal development but declines starting at postnatal day (PND) 10 and is virtually absent in the adult cortex. The mechanisms controlling this regulation are largely unknown. Methylation is important for gene silencing during development in many tissues, including the brain. In the present study, we examined the methylation status of ER alpha 5' untranslated exons during early postnatal development in male and female mice using methylation-specific PCR and pyrosequencing. Several regions of ER alpha promoter displayed a significant increase in methylation at PND 18 and 25 compared with PND 4. DNA methyltransferases (DNMT) are important for the initiation and maintenance of methylation. Real-time PCR showed that DNMT3A, the de novo DNMT peaked at PND 10 and was decreased by PND 25. DNMT1, which is important for maintenance of methylation, increased across development and stayed high in adult cortex. The methyl-CpG-binding protein 2 (MeCP2) is also important for stabilization of methylation. A chromatin immunoprecipitation assay showed a correlation between association of MeCP2 with ER alpha promoter and the increase in methylation and decrease in ER alpha expression after PND 10. In mice containing a mutant MeCP2 protein, ER alpha mRNA expression and promoter methylation patterns across development were different compared with wild-type mice. These data suggest that methylation of ER alpha promoters regulates ER alpha mRNA expression in the cortex during postnatal development in a MeCP2-dependent fashion.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Anti-MLL/HRX Antibody, NT., clone N4.4, clone N4.4, Upstate®, from mouse